missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. B23435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23435G 5 g
B23431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23435G Supplier TCI America Supplier No. B23435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72256-93-0
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.54
MDL Number MFCD03094696
InChI Key JRNDXUKMWFVEIC-UHFFFAOYSA-N
Synonym 4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
PubChem CID 2737503
IUPAC Name 4-bromo-3-methylbenzene-1-sulfonyl chloride
SMILES CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 59°C
Quantity 5 g
UN Number 3261
Formula Weight 269.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylbenzenesulfonyl Chloride