missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7123192
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38351G 1 g
B38355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38351G Supplier TCI America Supplier No. B38351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 351003-51-5
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD03094150
InChI Key IZCXVIACMHTZNA-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorobenzenesulphonyl chloride, 4-bromo-3-fluorobenzene-1-sulfonyl chloride, 3-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-3-fluorobenzenesulfonylchloride, 4-bromo-3-fluoro-benzenesulfonyl chloride, buttpark 76\07-90, benzenesulfonyl chloride, 4-bromo-3-fluoro, pubchem2220, acmc-209vnt, 4-bromo-3-fluorobenzene sulfonyl chloride
PubChem CID 2778218
IUPAC Name 4-bromo-3-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)F)Br

Specifications

Color Yellow
Boiling Point 218°C
Quantity 1 g
UN Number 3265
Formula Weight 273.50
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorobenzenesulfonyl Chloride