Learn More
4-Bromo-3-chloro-2-fluoropyridine, 98%
CAS: 1017793-21-3 | C5H2BrClFN | 210.43 g/mol
Supplier: Thermo Scientific Chemicals H6436203
Description
Useful intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUseful intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
| 4-Bromo-3-chloro-2-fluoropyridine | |
| C5H2BrClFN | |
| 1 g | |
| Slightly soluble in water. | |
| C1=CN=C(C(=C1Br)Cl)F | |
| 210.43 | |
| 210.43 | |
| Solid |
| 1017793-21-3 | |
| MFCD11040607 | |
| 4-bromo-3-chloro-2-fluoro-pyridine, pubchem22991, abbypharma ap-30-7506 | |
| UMQBGWKWOLRROL-UHFFFAOYSA-N | |
| 4-bromo-3-chloro-2-fluoropyridine | |
| 56604170 | |
| 98% |
Chemical Identifiers
| 1017793-21-3 | |
| 210.43 | |
| UMQBGWKWOLRROL-UHFFFAOYSA-N | |
| 56604170 | |
| C1=CN=C(C(=C1Br)Cl)F |
| C5H2BrClFN | |
| MFCD11040607 | |
| 4-bromo-3-chloro-2-fluoro-pyridine, pubchem22991, abbypharma ap-30-7506 | |
| 4-bromo-3-chloro-2-fluoropyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only