missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylisoxazole 98.0+%, TCI America™

Catalog No. p-7122999
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35191G 1 g
B35195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35191G Supplier TCI America Supplier No. B35191G
Only null left

Chemical Identifiers

CAS 10558-25-5
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD00068187
InChI Key GYHZPSUAMYIFQD-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
PubChem CID 318421
IUPAC Name 4-bromo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)Br

Specifications

Color Yellow
Boiling Point 75°C
Quantity 1 g
Formula Weight 176.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3,5-dimethylisoxazole
TSCA No
Recommended Storage Refrigerator