missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,5-dimethylisoxazole, 97%
CAS: 10558-25-5 | C5H6BrNO | 176.013 g/mol
Supplier: Thermo Scientific Chemicals A1417914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Bromo-3,5-dimethylisoxazole | |
| 1.49 | |
| 73°C (163°F) | |
| 1.488 | |
| 25 g | |
| Light sensitive | |
| GYHZPSUAMYIFQD-UHFFFAOYSA-N | |
| 4-bromo-3,5-dimethyl-1,2-oxazole | |
| 318421 | |
| 97% |
| 10558-25-5 | |
| 176°C | |
| C5H6BrNO | |
| MFCD00068187 | |
| 108522 | |
| 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n | |
| CC1=C(C(=NO1)C)Br | |
| 176.013 | |
| 176.02 |
Chemical Identifiers
| 10558-25-5 | |
| 176.013 | |
| GYHZPSUAMYIFQD-UHFFFAOYSA-N | |
| 318421 | |
| CC1=C(C(=NO1)C)Br |
| C5H6BrNO | |
| MFCD00068187 | |
| 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n | |
| 4-bromo-3,5-dimethyl-1,2-oxazole |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only