missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylisoxazole, 97%

Catalog No. AAA1417903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417903 1 g
AAA1417906 5 g
AAA1417914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1417903 Supplier Thermo Scientific Chemicals Supplier No. A1417903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10558-25-5 | C5H6BrNO | 176.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10558-25-5
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD00068187
InChI Key GYHZPSUAMYIFQD-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
PubChem CID 318421
IUPAC Name 4-bromo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)Br

Specifications

Density 1.49
Boiling Point 176°C
Flash Point 73°C (163°F)
Refractive Index 1.488
Quantity 1 g
Beilstein 108522
Sensitivity Light sensitive
Formula Weight 176.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-3,5-dimethylisoxazole
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only