missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-thiophenemethanol 98.0+%, TCI America™

Catalog No. B50601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50601G 1 g
B50605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50601G Supplier TCI America Supplier No. B50601G

Chemical Identifiers

CAS 79757-77-0
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.058
MDL Number MFCD04115392
InChI Key PXZNJHHUYJRFPZ-UHFFFAOYSA-N
PubChem CID 2795484
IUPAC Name (4-bromothiophen-2-yl)methanol
SMILES C1=C(SC=C1Br)CO

Specifications

Color White-Yellow
Quantity 1 g
Synonym 4-bromothiophen-2-yl methanol, 4-bromo-2-thienyl methanol, 4-bromo-2-hydroxymethylthiophene, 4-bromo-2-thiophenemethanol, 4-bromo-2-hydroxymethyl thiophene, 2-thiophenemethanol, 4-bromo, acmc-1bm1t, 3-bromo-5-thienyl methanol, 4-bromo-thiophene-2-methanol, 2-thiophenemethanol,4-bromo
Formula Weight 193.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2-thiophenemethanol