missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-thiophenecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7122916
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33865G 5 g
B338625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33865G Supplier TCI America Supplier No. B33865G
Only null left

Chemical Identifiers

CAS 16694-18-1
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
MDL Number MFCD03422294
InChI Key HJZFPRVFLBBAMU-UHFFFAOYSA-N
Synonym 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid
PubChem CID 610409
IUPAC Name 4-bromothiophene-2-carboxylic acid
SMILES C1=C(SC=C1Br)C(=O)O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 207.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-thiophenecarboxylic Acid