missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-thiophenecarboxylic acid, 97%, Thermo Scientific™
$28.59 - $28.59
Chemical Identifiers
| CAS | 16694-18-1 |
|---|---|
| Molecular Formula | C5H3BrO2S |
| Molecular Weight (g/mol) | 207.041 |
| InChI Key | HJZFPRVFLBBAMU-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid |
| PubChem CID | 610409 |
| IUPAC Name | 4-bromothiophene-2-carboxylic acid |
| SMILES | C1=C(SC=C1Br)C(=O)O |
Chemical Identifiers
| 16694-18-1 | |
| 207.041 | |
| 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid | |
| 4-bromothiophene-2-carboxylic acid |
| C5H3BrO2S | |
| HJZFPRVFLBBAMU-UHFFFAOYSA-N | |
| 610409 | |
| C1=C(SC=C1Br)C(=O)O |
Specifications
| 16694-18-1 | |
| Amber Glass Bottle | |
| 1 g | |
| HJZFPRVFLBBAMU-UHFFFAOYSA-N | |
| 4-bromothiophene-2-carboxylic acid | |
| 610409 | |
| 95% |
| 95% | |
| C5H3BrO2S | |
| 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid | |
| C1=C(SC=C1Br)C(=O)O | |
| 207.041 | |
| 207.05 | |
| 4-Bromo-2-thiophenecarboxylic acid |