missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. B40411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40411G 1 g
B40415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40411G Supplier TCI America Supplier No. B40411G

Chemical Identifiers

CAS 30766-03-1
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD03426665
InChI Key RPHHYRNGCJYQSP-UHFFFAOYSA-N
Synonym 4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
PubChem CID 735150
IUPAC Name 4-bromopyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1Br)C(=O)O

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 1 g
Formula Weight 202.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-pyridinecarboxylic Acid