missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7119904
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30335G 5 g
B303325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30335G Supplier TCI America Supplier No. B30335G
Only null left

Chemical Identifiers

CAS 875-51-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD00041312
InChI Key ZCWBZRBJSPWUPG-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-2-nitro, 2-nitro-4-bromoaniline, 4-bromo-o-nitroaniline, p-bromo-o-nitroaniline, 4-bromo-2-nitro-phenylamine, 4-bromo-2-nitrobenzeneamine, aniline, 4-bromo-2-nitro, 4-bromo-2-nitrobenzenamine, 1-amino-4-bromo-2-nitrobenzene, 4-bromo-2-nitro-aniline
PubChem CID 70132
IUPAC Name 4-bromo-2-nitroaniline
SMILES NC1=CC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitroaniline
RTECSNumber BW9370000
TSCA Yes