missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7122848
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32791G 1 g
B32795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32791G Supplier TCI America Supplier No. B32791G
Only null left

Chemical Identifiers

CAS 22282-99-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD03788196
InChI Key JFBMFWHEXBLFCR-UHFFFAOYSA-N
Synonym 4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine
PubChem CID 2762835
IUPAC Name 4-bromo-2-methylpyridine
SMILES CC1=CC(Br)=CC=N1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-methylpyridine