missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-hydroxypyridine, 97%

Catalog No. p-7031110
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH3206703 1 g
AAH3206706 5 g
2 options

Catalog No. AAH3206703

Supplier: Thermo Scientific Chemicals H3206703

Only null left

CAS: 36953-37-4 | C5H4BrNO | 173.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36953-37-4
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234041
InChI Key SSLMGOKTIUIZLY-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxypyridine, 4-bromopyridin-2-ol, 4-bromopyridin-2 1h-one, 4-bromo-2-pyridinol, 4-bromo-1,2-dihydropyridin-2-one, 4-bromo-2 1h-pyridinone, 4-bromo-2-pyridone, 4-bromo-2-hydroxy pyridine, 2-hydroxy-4-bromopyridine, 2 1h-pyridinone, 4-bromo
PubChem CID 817115
IUPAC Name 4-bromo-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1Br

Specifications

Melting Point 201°C to 204°C
Quantity 1 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-hydroxypyridine
Hazard Category H302+H332-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280g-P280i-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only