missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-furaldehyde 97.0+%, TCI America™

Catalog No. B18161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18161G 1 g
B18165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18161G Supplier TCI America Supplier No. B18161G
Only null left

Chemical Identifiers

CAS 21921-76-6
Molecular Formula C5H3BrO2
Molecular Weight (g/mol) 174.981
MDL Number MFCD00671504
InChI Key MRGBBKQOSUHKPF-UHFFFAOYSA-N
Synonym 4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi
PubChem CID 2757010
IUPAC Name 4-bromofuran-2-carbaldehyde
SMILES C1=C(OC=C1Br)C=O

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.98
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-furaldehyde
TSCA No
Recommended Storage Refrigerator