missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7123108
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B36985G 5 g
B369825G 25 g
2 options

Catalog No. B36985G

Supplier: TCI America B36985G

Only null left

Chemical Identifiers

CAS 216159-03-4
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD01090993
InChI Key XNYBZLRIUHNRQY-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzene-1-sulfonyl chloride, 2-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-2-fluorobenzenesulphonyl chloride, 4-bromo-2-fluoro-benzenesulfonyl chloride, buttpark 147\11-41, 4-bromo-2-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 4-bromo-2-fluoro, pubchem5065, acmc-209via, ksc201o8l
PubChem CID 2736310
IUPAC Name 4-bromo-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Br)F)S(=O)(=O)Cl

Specifications

Melting Point 66°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 273.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzenesulfonyl Chloride