missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzenesulfonyl chloride, 98%

Catalog No. p-7027529
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2435206 5 g
AAB2435214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2435206 Supplier Thermo Scientific Chemicals Supplier No. B2435206
Only null left

CAS: 216159-03-4 | C6H3BrClFO2S | 273.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216159-03-4
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD01090993
InChI Key XNYBZLRIUHNRQY-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzene-1-sulfonyl chloride, 2-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-2-fluorobenzenesulphonyl chloride, 4-bromo-2-fluoro-benzenesulfonyl chloride, buttpark 147\11-41, 4-bromo-2-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 4-bromo-2-fluoro, pubchem5065, acmc-209via, ksc201o8l
PubChem CID 2736310
IUPAC Name 4-bromo-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Br)F)S(=O)(=O)Cl

Specifications

Melting Point 63°C to 66°C
Boiling Point 84°C (0.5 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 273.51
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only