missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-fluoroacetophenone, 96%

Catalog No. AAH6212703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6212703 1 g
AAH6212706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6212703 Supplier Thermo Scientific Chemicals Supplier No. H6212703
Only null left

CAS: 625446-22-2 | C8H6BrFO | 217.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625446-22-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD03411551
InChI Key ASKFCSCYGAFWAB-UHFFFAOYSA-N
Synonym 1-4-bromo-2-fluorophenyl ethanone, 4-bromo-2-fluoroacetophenone, 4'-bromo-2'-fluoroacetophenone, 1-4-bromo-2-fluorophenyl ethan-1-one, 1-acetyl-4-bromo-2-fluorobenzene, ethanone, 1-4-bromo-2-fluorophenyl, 2-fluoro-4-bromoacetophenone, 1-4-bromo-2-fluoro-phenyl-ethanone, acmc-209n4s, ksc352o4j
PubChem CID 21938581
IUPAC Name 1-(4-bromo-2-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Br)F

Specifications

Quantity 1 g
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4'-Bromo-2'-fluoroacetophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only