missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog No. p-7058179
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64753MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH64753MD Supplier Thermo Scientific Chemicals Supplier No. H64753MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an primary and secondary intermediate for pharmaceutical, organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 885702-34-1
Molecular Formula C4H2BrClN2
Molecular Weight (g/mol) 193.43
MDL Number MFCD08272196
InChI Key FBEBVAQOMVWORE-UHFFFAOYSA-N
Synonym pyrimidine, 4-bromo-2-chloro-9ci, pyrimidine, 4-bromo-2-chloro, 2-chloro-4-bromopyrimidine
PubChem CID 45117394
IUPAC Name 4-bromo-2-chloropyrimidine
SMILES ClC1=NC=CC(Br)=N1

Specifications

Quantity 250 mg
Beilstein 10334333
Solubility Information Slightly soluble in water.
Formula Weight 193.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chloropyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only