missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyridine, 97%

Catalog No. AC434800050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434800050 5 g Glass bottle
AC434800250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434800050 Supplier Thermo Scientific Chemicals Supplier No. 434800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73583-37-6 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73583-37-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD03840756
InChI Key ONHMWUXYIFULDO-UHFFFAOYSA-N
Synonym 2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
PubChem CID 3847770
IUPAC Name 4-bromo-2-chloropyridine
SMILES C1=CN=C(C=C1Br)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.7340g/mL
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.734
Formula Weight 192.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-2-chloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only