missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyridine, 94%

Catalog No. AAH2729906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2729906 5 g
AAH2729914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2729906 Supplier Thermo Scientific Chemicals Supplier No. H2729906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 73583-37-6 | C5H3BrClN | 192.44 g/mol

It is used in the synthesis of quaterpyridine nemertelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of quaterpyridine nemertelline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 73583-37-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD03840756
InChI Key ONHMWUXYIFULDO-UHFFFAOYSA-N
Synonym 2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
PubChem CID 3847770
IUPAC Name 4-bromo-2-chloropyridine
SMILES C1=CN=C(C=C1Br)Cl

Specifications

Melting Point 18°C to 20°C
Density 1.734
Boiling Point 70°C (3 mmHg)
Flash Point 104°C (219°F)
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 192.44
Percent Purity 94%
Chemical Name or Material 4-Bromo-2-chloropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only