missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzenesulfonyl chloride, 96%

Catalog No. p-7027861
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2491306 5 g
AAB2491314 25 g
AAB2491322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2491306 Supplier Thermo Scientific Chemicals Supplier No. B2491306

CAS: 351003-52-6 | C6H3BrCl2O2S | 289.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351003-52-6
Molecular Formula C6H3BrCl2O2S
Molecular Weight (g/mol) 289.95
MDL Number MFCD03094630
InChI Key OECYEHWJPRPCOH-UHFFFAOYSA-N
Synonym 4-bromo-2-chlorobenzene-1-sulfonyl chloride, 2-chloro-4-bromo-benzenesulfonyl chloride, 4-bromo-2-chlorobenzenesulphonyl chloride, 4-bromo-2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 4-bromo-2-chloro, 4-bromo-2-chlorophenylsulfonyl chloride, pubchem11732, 4-bromo-2-chloro-benzenesulfonyl chloride, acmc-1ag54, ksc221s9p
PubChem CID 2734766
IUPAC Name 4-bromo-2-chlorobenzenesulfonyl chloride
SMILES ClC1=CC(Br)=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 76°C to 81°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 289.96
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only