missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-chloroacetanilide, 98+%

Catalog No. p-7024281
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1865214 25 g
AAA1865222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1865214 Supplier Thermo Scientific Chemicals Supplier No. A1865214

CAS: 3460-23-9 | C8H7BrClNO | 248.504 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Soluble in hot methanol (very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 3460-23-9
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040852
InChI Key MITWNEIUIPGZKR-UHFFFAOYSA-N
Synonym 4'-bromo-2'-chloroacetanilide, n-4-bromo-2-chlorophenyl acetamide, 4-bromo-2-chloroacetanilide, acetamide, n-4-bromo-2-chlorophenyl, maybridge3_007422, 2-chlor-4-bromacetanilid, acmc-209i8u, buttpark 83\07-60, acetanilide, 4'-bromo-2'-chloro
PubChem CID 610167
IUPAC Name N-(4-bromo-2-chlorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Br)Cl

Specifications

Melting Point 151°C to 152°C
Quantity 25 g
Beilstein 2719133
Solubility Information Soluble in hot methanol (very faint turbidity).
Formula Weight 248.51
Percent Purity ≥98%
Chemical Name or Material 4'-Bromo-2'-chloroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only