missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(5-isoxazolyl)phenol, 97%

Catalog No. AAH2755714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2755714 25 g
AAH2755703 1 g
AAH2755706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2755714 Supplier Thermo Scientific Chemicals Supplier No. H2755714
Only null left

CAS: 213690-27-8 | C9H6BrNO2 | 240.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 213690-27-8
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.06
MDL Number MFCD00799493
InChI Key GBXOJTWVFYSJRI-UHFFFAOYSA-N
Synonym 4-bromo-2-5-isoxazolyl phenol, 4-bromo-6-2h-1,2-oxazol-5-ylidene cyclohexa-2,4-dien-1-one, 4-bromo-2-1,2-oxazol-5-yl phenol, acmc-1ckhc, 4-bromo-2-isoxazole-5-yl phenol, phenol,4-bromo-2-5-isoxazolyl
PubChem CID 6744794
IUPAC Name 4-bromo-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES BrC1=CC(=C2ONC=C2)C(=O)C=C1

Specifications

Melting Point 207°C to 212°C
Quantity 25 g
Formula Weight 240.05
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(5-isoxazolyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only