missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-difluoro-1,3-benzodioxole, 97%

Catalog No. p-7027550
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2438903 1 g
AAB2438906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2438903 Supplier Thermo Scientific Chemicals Supplier No. B2438903

CAS: 144584-66-7 | C7H3BrF2O2 | 237 g/mol

4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is unstable under normal temperature. storage temperature is 2 - 4°C. Incompatible material is oxidizing agents.

Chemical Identifiers

CAS 144584-66-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD01631385
InChI Key LSZYHXNOLVSZHH-UHFFFAOYSA-N
Synonym 4-bromo-2,2-difluorobenzo d 1,3 dioxole, 4-bromo-2,2-difluoro-2h-1,3-benzodioxole, 2,2-difluoro-4-bromo-1,3-benzodioxole, 1,3-benzodioxol-4-bromo, 2,2-difluoro, 4-bromo-2,2-difluoro-benzo 1,3 dioxole, 4-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-20a3ue, 4-bromo-2,2-difluoro-1,3-benzodioxol, 1,3-benzodioxole,4-bromo-2,2-difluoro, 4-bromo-2,2-difluoro-1,3-benzodioxolane
PubChem CID 2773297
IUPAC Name 4-bromo-2,2-difluoro-1,3-benzodioxole
SMILES C1=CC2=C(C(=C1)Br)OC(O2)(F)F

Specifications

Refractive Index 1.4968
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,2-difluoro-1,3-benzodioxole
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only