missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromo-2,2':6',2″-terpyridine 97.0+%, TCI America™
$976.76 - $2615.27
Chemical Identifiers
| CAS | 149817-62-9 |
|---|---|
| Molecular Formula | C15H10BrN3 |
| Molecular Weight (g/mol) | 312.17 |
| MDL Number | MFCD06796965 |
| InChI Key | CJRRILXBSCRHKN-UHFFFAOYSA-N |
| PubChem CID | 10614995 |
| IUPAC Name | 4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine |
| SMILES | BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 |
Chemical Identifiers
| 149817-62-9 | |
| 312.17 | |
| CJRRILXBSCRHKN-UHFFFAOYSA-N | |
| 4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine |
| C15H10BrN3 | |
| MFCD06796965 | |
| 10614995 | |
| BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 |
Specifications
| 149817-62-9 | |
| White-Yellow | |
| MFCD06796965 | |
| 4′C-Bromo-alpha,alpha′C,alpha′C′C-tripyridyl | |
| BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1 | |
| 312.17 | |
| 312.17 | |
| Crystalline Powder |
| 139°C | |
| C15H10BrN3 | |
| 1 g | |
| CJRRILXBSCRHKN-UHFFFAOYSA-N | |
| 4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine | |
| 10614995 | |
| ≥97.0% (HPLC,T) | |
| 4′-Bromo-2,2′:6′,2′′-terpyridine |
Safety and Handling
TSCA : No