Learn More
4'-Bromo-2,2,2-trifluoroacetophenone, 97%
CAS: 16184-89-7 | C8H4BrF3O | 253.018 g/mol
$250.67 - $787.33
Chemical Identifiers
| CAS | 16184-89-7 |
|---|---|
| Molecular Formula | C8H4BrF3O |
| Molecular Weight (g/mol) | 253.018 |
| MDL Number | MFCD00191862 |
| InChI Key | IHGSAQHSAGRWNI-UHFFFAOYSA-N |
| Synonym | 4'-bromo-2,2,2-trifluoroacetophenone, 1-4-bromophenyl-2,2,2-trifluoroethanone, 1-4-bromophenyl-2,2,2-trifluoroethan-1-one, 1-4-bromo-phenyl-2,2,2-trifluoro-ethanone, ethanone, 1-4-bromophenyl-2,2,2-trifluoro, 1-4-bromophenyl-2,2,2-trifluoro-1-ethanone, intermediates-zcf02216, 4-trifluoroacetylbromobenzene, 4'-bromo-2,2,2-trifluoracetophenone, 4'-bromo-2,2,2-trifluoroaceto-phenone |
| PubChem CID | 603866 |
| IUPAC Name | 1-(4-bromophenyl)-2,2,2-trifluoroethanone |
| SMILES | C1=CC(=CC=C1C(=O)C(F)(F)F)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3362703
|
Thermo Scientific Chemicals
H3362703 |
1 g |
Each for $250.67
|
|
|||||
|
AAH3362706
|
Thermo Scientific Chemicals
H3362706 |
5 g |
Each for $787.33
|
|
|||||
Description
4?-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
Chemical Identifiers
| 16184-89-7 | |
| 253.018 | |
| IHGSAQHSAGRWNI-UHFFFAOYSA-N | |
| 603866 | |
| C1=CC(=CC=C1C(=O)C(F)(F)F)Br |
| C8H4BrF3O | |
| MFCD00191862 | |
| 4'-bromo-2,2,2-trifluoroacetophenone, 1-4-bromophenyl-2,2,2-trifluoroethanone, 1-4-bromophenyl-2,2,2-trifluoroethan-1-one, 1-4-bromo-phenyl-2,2,2-trifluoro-ethanone, ethanone, 1-4-bromophenyl-2,2,2-trifluoro, 1-4-bromophenyl-2,2,2-trifluoro-1-ethanone, intermediates-zcf02216, 4-trifluoroacetylbromobenzene, 4'-bromo-2,2,2-trifluoracetophenone, 4'-bromo-2,2,2-trifluoroaceto-phenone | |
| 1-(4-bromophenyl)-2,2,2-trifluoroethanone |
Specifications
| 16184-89-7 | |
| 95°C (4 mmHg) | |
| MFCD00191862 | |
| Moisture sensitive | |
| Slightly soluble in water. | |
| C1=CC(=CC=C1C(=O)C(F)(F)F)Br | |
| 253.018 | |
| 253.02 | |
| 4'-Bromo-2,2,2-trifluoroacetophenone |
| 26°C to 30°C | |
| C8H4BrF3O | |
| 1 g | |
| 4'-bromo-2,2,2-trifluoroacetophenone, 1-4-bromophenyl-2,2,2-trifluoroethanone, 1-4-bromophenyl-2,2,2-trifluoroethan-1-one, 1-4-bromo-phenyl-2,2,2-trifluoro-ethanone, ethanone, 1-4-bromophenyl-2,2,2-trifluoro, 1-4-bromophenyl-2,2,2-trifluoro-1-ethanone, intermediates-zcf02216, 4-trifluoroacetylbromobenzene, 4'-bromo-2,2,2-trifluoracetophenone, 4'-bromo-2,2,2-trifluoroaceto-phenone | |
| IHGSAQHSAGRWNI-UHFFFAOYSA-N | |
| 1-(4-bromophenyl)-2,2,2-trifluoroethanone | |
| 603866 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only