missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2,2,2-trifluoroacetophenone, 97%

Catalog No. AAH3362703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3362703 1 g
AAH3362706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3362703 Supplier Thermo Scientific Chemicals Supplier No. H3362703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16184-89-7 | C8H4BrF3O | 253.018 g/mol

4?-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.

Chemical Identifiers

CAS 16184-89-7
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.018
MDL Number MFCD00191862
InChI Key IHGSAQHSAGRWNI-UHFFFAOYSA-N
Synonym 4'-bromo-2,2,2-trifluoroacetophenone, 1-4-bromophenyl-2,2,2-trifluoroethanone, 1-4-bromophenyl-2,2,2-trifluoroethan-1-one, 1-4-bromo-phenyl-2,2,2-trifluoro-ethanone, ethanone, 1-4-bromophenyl-2,2,2-trifluoro, 1-4-bromophenyl-2,2,2-trifluoro-1-ethanone, intermediates-zcf02216, 4-trifluoroacetylbromobenzene, 4'-bromo-2,2,2-trifluoracetophenone, 4'-bromo-2,2,2-trifluoroaceto-phenone
PubChem CID 603866
IUPAC Name 1-(4-bromophenyl)-2,2,2-trifluoroethanone
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)Br

Specifications

Melting Point 26°C to 30°C
Boiling Point 95°C (4 mmHg)
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 253.02
Percent Purity 97%
Chemical Name or Material 4'-Bromo-2,2,2-trifluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only