missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-indazole, 97+%

Catalog No. AAH6452706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6452706 5 g
AAH6452714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6452706 Supplier Thermo Scientific Chemicals Supplier No. H6452706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 186407-74-9 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 186407-74-9
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD05664001
InChI Key KJIODOACRIRBPB-UHFFFAOYSA-N
Synonym 4-bromoindazole, 1h-indazole, 4-bromo, 4-bromo 1h indazole, 4-bromo-indazole, bromo-1h-indazole, pubchem7834, 4-bromo-2h-indazole, acmc-1bzds, 4-bromanyl-1h-indazole
PubChem CID 22352548
IUPAC Name 4-bromo-1H-indazole
SMILES C1=CC2=C(C=NN2)C(=C1)Br

Specifications

Melting Point 164°C to 169°C
Color White
Quantity 5 g
UN Number UN2811
Beilstein 9757929
Formula Weight 197.04
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 4-Bromo-1H-indazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only