missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-methylpyrazole 98.0+%, TCI America™
Supplier: TCI America B40605G
Specifications
| 4-Bromo-1-methylpyrazole | |
| Yellow | |
| C4H5BrN2 | |
| 5 g | |
| IXJSDKIJPVSPKF-UHFFFAOYSA-N | |
| 4-bromo-1-methyl-1H-pyrazole | |
| 167433 | |
| ≥98.0% (GC) |
| 15803-02-8 | |
| 78°C | |
| MFCD02179565 | |
| 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp | |
| CN1C=C(Br)C=N1 | |
| 161.00 | |
| 161.00 | |
| Liquid |
Chemical Identifiers
| 15803-02-8 | |
| 161.00 | |
| IXJSDKIJPVSPKF-UHFFFAOYSA-N | |
| 167433 | |
| CN1C=C(Br)C=N1 |
| C4H5BrN2 | |
| MFCD02179565 | |
| 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp | |
| 4-bromo-1-methyl-1H-pyrazole |
Safety and Handling
TSCA : No