Learn More
4-Bromo-1-methyl-1H-pyrazole, 98+%
CAS: 15803-02-8 | C4H5BrN2 | 161.00 g/mol
$119.16 - $451.92
Chemical Identifiers
| CAS | 15803-02-8 |
|---|---|
| Molecular Formula | C4H5BrN2 |
| Molecular Weight (g/mol) | 161.00 |
| MDL Number | MFCD02179565 |
| InChI Key | IXJSDKIJPVSPKF-UHFFFAOYSA-N |
| Synonym | 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp |
| PubChem CID | 167433 |
| IUPAC Name | 4-bromo-1-methylpyrazole |
| SMILES | CN1C=C(Br)C=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6476814
|
Thermo Scientific Chemicals
H6476814 |
25 g |
Each for $119.16
|
|
|||||
|
AAH6476822
|
Thermo Scientific Chemicals
H6476822 |
100 g |
Each for $451.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15803-02-8 | |
| 161.00 | |
| IXJSDKIJPVSPKF-UHFFFAOYSA-N | |
| 167433 | |
| CN1C=C(Br)C=N1 |
| C4H5BrN2 | |
| MFCD02179565 | |
| 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp | |
| 4-bromo-1-methylpyrazole |
Specifications
| 15803-02-8 | |
| 1.558 g/mL | |
| 94°C (201°F) | |
| 1.531 | |
| 25 g | |
| 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp | |
| CN1C=C(Br)C=N1 | |
| 161.00 | |
| 161 | |
| Liquid |
| Colorless to Yellow | |
| 185°C to 188°C | |
| C4H5BrN2 | |
| MFCD02179565 | |
| 108120 | |
| IXJSDKIJPVSPKF-UHFFFAOYSA-N | |
| 4-bromo-1-methylpyrazole | |
| 167433 | |
| ≥98% | |
| 4-Bromo-1-methyl-1H-pyrazole |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only