missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-1-méthyl-1H-pyrazol-3-yl)méthanol, ≥97%, Thermo Scientific™

Numéro de catalogue. CC63909DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC63909DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC63909DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC63909DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 915707-65-2
Formule moléculaire C5H7BrN2O
Poids moléculaire (g/mol) 191.028
Numéro MDL MFCD06202888
Clé InChI JFGLJTFTVBZOCB-UHFFFAOYSA-N
Synonyme 4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
PubChem CID 24229713
Nom de l’IUPAC (4-bromo-1-méthylpyrazol-3-yl)méthanol
SOURIRES CN1C=C(C(=N1)CO)Br

Spécifications

Nom ou substance chimique (4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97+%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 191.03
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.