missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-imidazole, 95%

Catalog No. AC452430010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452430010 1 g
AC452430050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452430010 Supplier Thermo Scientific Chemicals Supplier No. 452430010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25676-75-9 | C4H5BrN2 | 161.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25676-75-9
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD01320501
InChI Key IOTSLMMLLXTNNH-UHFFFAOYSA-N
PubChem CID 1277653
IUPAC Name 4-bromo-1-methylimidazole
SMILES CN1C=NC(Br)=C1

Specifications

Boiling Point 91°C to 93°C (0.1 hPa)
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 4-bromo-1-methyl-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, acmc-1cjr4, 4-bromo-1-methyl-imidazole, 4-bromanyl-1-methyl-imidazole, 1-methyl-4-bromo-1h-imidazole, 1h-imidazole,4-bromo-1-methyl
Formula Weight 161
Percent Purity 95%
Chemical Name or Material 4-Bromo-1-methyl-1H-imidazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only