missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-chloro-2-ethylbenzene, 98+%

Catalog No. AAB2561314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2561314 25 g
AAB2561306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2561314 Supplier Thermo Scientific Chemicals Supplier No. B2561314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 289039-22-1 | C8H8BrCl | 219.506 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 289039-22-1
Molecular Formula C8H8BrCl
Molecular Weight (g/mol) 219.506
MDL Number MFCD00672954
InChI Key ILQNJGBEXZUYOI-UHFFFAOYSA-N
Synonym 5-bromo-2-chloroethylbenzene, acmc-1cnam, 4-bromo-1-chloro-2-ethyl benzene, 4-bromo-1-chloro-2-ethyl-benzene
PubChem CID 44119401
IUPAC Name 4-bromo-1-chloro-2-ethylbenzene
SMILES CCC1=C(C=CC(=C1)Br)Cl

Specifications

Quantity 25 g
Formula Weight 219.51
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-1-chloro-2-ethylbenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only