missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-hydroxypyridine, 97%

Catalog No. AC432090050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432090050 5 g Glass bottle
AC432090010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432090050 Supplier Thermo Scientific Chemicals Supplier No. 432090050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 36953-37-4 | C5H4BrNO | 174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36953-37-4
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174
MDL Number MFCD00234041
InChI Key SSLMGOKTIUIZLY-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxypyridine, 4-bromopyridin-2-ol, 4-bromopyridin-2 1h-one, 4-bromo-2-pyridinol, 4-bromo-1,2-dihydropyridin-2-one, 4-bromo-2 1h-pyridinone, 4-bromo-2-pyridone, 4-bromo-2-hydroxy pyridine, 2-hydroxy-4-bromopyridine, 2 1h-pyridinone, 4-bromo
PubChem CID 817115
IUPAC Name 4-bromo-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 203°C to 208°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 174
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-hydroxypyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only