missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,3,5-trimethylpyrazole 98.0+%, TCI America™

Catalog No. B47441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47441G 1 g
B47445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47441G Supplier TCI America Supplier No. B47441G
Only null left

Chemical Identifiers

CAS 15801-69-1
Molecular Formula C6H9BrN2
Molecular Weight (g/mol) 189.056
MDL Number MFCD00052532
InChI Key UNTQXOJGXGRHMG-UHFFFAOYSA-N
Synonym 4-bromo-1,3,5-trimethyl-1h-pyrazole, 4-bromo-2,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-2h-pyrazole, pubchem20507, acmc-209dhc, buttpark 34\09-42, 4-bromo-1,3,5-trimethylpyrazole, 4-bromo-1,3,5-trimethyl-pyrazole, 4-bromo-1,3,5-trimethyl pyrazole
PubChem CID 519181
IUPAC Name 4-bromo-1,3,5-trimethylpyrazole
SMILES CC1=C(C(=NN1C)C)Br

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 189.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-1,3,5-trimethylpyrazole