missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-(methylenedioxy)benzene, 97%

Catalog No. AC307680250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307680250 25 g Glass bottle
AC307681000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC307680250 Supplier Thermo Scientific Chemicals Supplier No. 307680250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2635-13-4 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2635-13-4
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00005821
InChI Key FBOYMIDCHINJKC-UHFFFAOYSA-N
Synonym 4-bromo-1,2-methylenedioxy benzene, 5-bromobenzo d 1,3 dioxole, 4-bromo-1,2-methylenedioxybenzene, 1-bromo-3,4-methylenedioxy benzene, 3,4-methylenedioxybromobenzene, 5-bromo-2h-1,3-benzodioxole, 5-bromobenzo-1,3-dioxole, 5-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo, 3,4-methylenedioxy bromobenzene
PubChem CID 75831
SMILES BrC1=CC=C2OCOC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange-Yellow
Density 1.6600g/mL
Boiling Point 85°C to 86°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5820 to 1.584
Quantity 25 g
Specific Gravity 1.66
IUPAC Name 5-bromo-2H-1,3-benzodioxole
Formula Weight 201.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1, 2-(methylenedioxy)benzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-123-5

RUO – Research Use Only