missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)benzeneboronic acid, 95%

Catalog No. p-7035508
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH53225MD 250 mg
AAH5322503 1 g
2 options

Catalog No. AAH53225MD

Supplier: Thermo Scientific Chemicals H53225MD

Only null left

CAS: 489446-42-6 | C12H18BNO4 | 251.09 g/mol

4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 489446-42-6
Molecular Formula C12H18BNO4
Molecular Weight (g/mol) 251.09
MDL Number MFCD04115637
InChI Key MUBGEKQUCSEECZ-UHFFFAOYSA-N
Synonym 4-n-boc-aminomethyl phenylboronic acid, 4-n-boc-amino methyl phenylboronic acid, 4-tert-butoxycarbonyl amino methyl phenyl boronic acid, 4-n-boc aminomethylphenylboronic acid, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 4-boc-aminomethyl benzeneboronic acid, 4-aminomethyl benzeneboronic acid, n-boc protected, 4-tert-butoxycarbonylamino methyl phenylboronic acid
PubChem CID 3863163
IUPAC Name [4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 204°C to 206°C
Quantity 250 mg
Formula Weight 251.09
Percent Purity 97%
Chemical Name or Material 4-(Boc-aminomethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only