missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)piperidine, 96%

CAS: 73874-95-0 | C10H21N2O2 | 201.29 g/mol

$95.49 - $955.03

Chemical Identifiers

CAS 73874-95-0
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD00798171
InChI Key CKXZPVPIDOJLLM-UHFFFAOYSA-O
Synonym 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
PubChem CID 723833
SMILES CC(C)(C)OC(=O)NC1CC[NH2+]CC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2851703
View Documents
Thermo Scientific Chemicals
H2851703
1 g
Each for $95.49
Only null left
 
AAH2851706
View Documents
Thermo Scientific Chemicals
H2851706
5 g
Each for $294.81
Only null left
 
AAH2851714
View Documents
Thermo Scientific Chemicals
H2851714
25 g
Each for $955.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

73874-95-0
201.29
CKXZPVPIDOJLLM-UHFFFAOYSA-O
723833
C10H21N2O2
MFCD00798171
4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
CC(C)(C)OC(=O)NC1CC[NH2+]CC1

Specifications

162°C to 166°C
C10H21N2O2
4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
CC(C)(C)OC(=O)NC1CC[NH2+]CC1
1 g
723833
96%
73874-95-0
MFCD00798171
CKXZPVPIDOJLLM-UHFFFAOYSA-O
tert-butyl N-piperidin-4-ylcarbamate
201.29
200.28
4-(Boc-amino)piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 616-026-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only