missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)piperidine, 96%

Catalog No. AAH2851703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2851703 1 g
AAH2851706 5 g
AAH2851714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2851703 Supplier Thermo Scientific Chemicals Supplier No. H2851703

CAS: 73874-95-0 | C10H21N2O2 | 201.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73874-95-0
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD00798171
InChI Key CKXZPVPIDOJLLM-UHFFFAOYSA-O
Synonym 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
PubChem CID 723833
SMILES CC(C)(C)OC(=O)NC1CC[NH2+]CC1

Specifications

Melting Point 162°C to 166°C
IUPAC Name tert-butyl N-piperidin-4-ylcarbamate
Quantity 1 g
Formula Weight 200.28
Percent Purity 96%
Chemical Name or Material 4-(Boc-amino)piperidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 616-026-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only