missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%

Catalog No. p-7052688
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5531103 1 g
AAH5531106 5 g
AAH5531114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5531103 Supplier Thermo Scientific Chemicals Supplier No. H5531103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 153747-97-8 | C14H20BrN3O2 | 342.237 g/mol

4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 153747-97-8
Molecular Formula C14H20BrN3O2
Molecular Weight (g/mol) 342.237
MDL Number MFCD07369772
InChI Key DSLVSFMWCDGZIL-UHFFFAOYSA-N
Synonym tert-butyl 4-5-bromopyridin-2-yl piperazine-1-carboxylate, 2-4-boc-piperazino-5-bromopyridine, 4-boc-1-5-bromo-2-pyridyl piperazine, 5-bromo-2-4-n-boc piperazin-1-yl pyridine, 4-5-bromo-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 5-bromo-2-4-tert-butoxycarbonylpiperazin-1-yl pyridine, 4-5-bromopyridin-2-yl piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-5-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ah70, 2-n-boc-piperazin-1-yl-5-bromopyridine
PubChem CID 11244775
IUPAC Name tert-butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)Br

Specifications

Melting Point 83°C to 87°C
Color White to Yellow
Quantity 1 g
UN Number UN2811
Solubility Information Soluble in water.
Formula Weight 342.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Boc-1-(5-bromo-2-pyridyl)piperazine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only