missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene 98.0+%, TCI America™
Supplier: TCI America B4796200MG
Specifications
| 4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene | |
| C42H35N3 | |
| 200 mg | |
| CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(C=C1)N=NC1=CC=CC=C1)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C)C | |
| 581.76 | |
| 581.76 | |
| Crystalline Powder |
| 883554-70-9 | |
| MFCD29089342 | |
| XIVVSHQOQWKLRF-UHFFFAOYSA-N | |
| N-(9,9-dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-[4-(2-phenyldiazen-1-yl)phenyl]-9H-fluoren-2-amine | |
| 91114977 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 883554-70-9 | |
| 581.76 | |
| XIVVSHQOQWKLRF-UHFFFAOYSA-N | |
| N-(9,9-dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-[4-(2-phenyldiazen-1-yl)phenyl]-9H-fluoren-2-amine |
| C42H35N3 | |
| MFCD29089342 | |
| 91114977 | |
| CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(C=C1)N=NC1=CC=CC=C1)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C)C |
Safety and Handling
TSCA : No