missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylyl(2,4,6-trimethoxyphenyl)iodonium Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. B5573200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5573200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5573200MG Supplier TCI America Supplier No. B5573200MG

Chemical Identifiers

CAS 1868173-47-0
Molecular Formula C22H20F3IO6S
Synonym 4-Biphenylyl(2,4,6-trimethoxyphenyl)iodonium Triflate, [1,1′-Biphenyl]-4-yl(2,4,6-trimethoxyphenyl)iodonium Trifluoromethanesulfonate

Specifications

Melting Point 135°C
Quantity 200 mg
Formula Weight 596.36
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylyl(2,4,6-trimethoxyphenyl)iodonium Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator