missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylsulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7119547
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21995G 5 g
B219925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21995G Supplier TCI America Supplier No. B21995G
Only null left

Chemical Identifiers

CAS 1623-93-4
Molecular Formula C12H9ClO2S
Molecular Weight (g/mol) 252.71
MDL Number MFCD00078658
InChI Key ALBQXDHCMLLQMB-UHFFFAOYSA-N
Synonym 4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990
PubChem CID 74192
IUPAC Name [1,1'-biphenyl]-4-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 114°C
Quantity 5 g
UN Number 3261
Formula Weight 252.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylsulfonyl Chloride