missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylsulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1623-93-4
Molecular Formula C12H9ClO2S
Molecular Weight (g/mol) 252.71
MDL Number MFCD00078658
InChI Key ALBQXDHCMLLQMB-UHFFFAOYSA-N
Synonym 4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990
PubChem CID 74192
IUPAC Name 4-phenylbenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391960050
SDS View Documents
Thermo Scientific Chemicals
391960050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC391960250
SDS View Documents
Thermo Scientific Chemicals
391960250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1623-93-4
252.71
ALBQXDHCMLLQMB-UHFFFAOYSA-N
74192
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
C12H9ClO2S
MFCD00078658
4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990
4-phenylbenzenesulfonyl chloride

Specifications

1623-93-4
100.0
96% min. (HPLC)
C12H9ClO2S
MFCD00078658
4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990
ALBQXDHCMLLQMB-UHFFFAOYSA-N
4-phenylbenzenesulfonyl chloride
74192
97%
98.0
Authentic
Glass bottle
C6H5C6H4SO2Cl
5g
Solubility in water: reacts. Other solubilities: soluble in common organic solvents
ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
252.71
252.72
4-Biphenylsulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 216-609-1

SDS
Documents

Documents

RUO – Research Use Only