missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylcarboxaldehyde, 99%

Catalog No. AC106260050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106260050 5 g Glass bottle
AC106260250 25 g Glass bottle
AC106261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106260050 Supplier Thermo Scientific Chemicals Supplier No. 106260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3218-36-8 | C13H10O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name 4-phenylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color White to Yellow
Boiling Point 184.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5C6H4CHO
Quantity 5 g
Beilstein 07, 430
Solubility Information Solubility in water: insoluble
Formula Weight 182.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Biphenylcarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 221-742-3

RUO – Research Use Only