Learn More
4-Biphenylcarbonyl chloride, 98%
CAS: 14002-51-8 | C13H9ClO | 216.66 g/mol
$113.91 - $113.91
Chemical Identifiers
| CAS | 14002-51-8 |
|---|---|
| Molecular Formula | C13H9ClO |
| Molecular Weight (g/mol) | 216.66 |
| MDL Number | MFCD00000692 |
| InChI Key | JPVUWCPKMYXOKW-UHFFFAOYSA-N |
| Synonym | 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride |
| PubChem CID | 84151 |
| IUPAC Name | 4-phenylbenzoyl chloride |
| SMILES | ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC171250100
|
Thermo Scientific Chemicals
171250100 |
10 g |
Each for $113.91
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14002-51-8 | |
| 216.66 | |
| JPVUWCPKMYXOKW-UHFFFAOYSA-N | |
| 84151 | |
| ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H9ClO | |
| MFCD00000692 | |
| 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride | |
| 4-phenylbenzoyl chloride |
Specifications
| 14002-51-8 | |
| 100.0 | |
| White to Yellow | |
| 97.5% min. (ex Cl) (Argentometry) | |
| C13H9ClO | |
| MFCD00000692 | |
| 09, I, 280 | |
| 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride | |
| JPVUWCPKMYXOKW-UHFFFAOYSA-N | |
| 4-phenylbenzoyl chloride | |
| 84151 | |
| 98% | |
| 4-Biphenylcarbonyl chloride, 98% |
| 97.5 | |
| 110°C to 114°C | |
| Authentic | |
| Glass bottle | |
| C6H5C6H4COCl | |
| 10 g | |
| 04,376; 07,282 | |
| Solubility in water: reacts. Other solubilities: soluble in chloroform,dmf | |
| ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
| 216.66 | |
| 216.67 | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 237-804-8
TSCA : TSCA
RUO – Research Use Only