missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$57.75 - $57.75
Chemical Identifiers
| CAS | 5122-94-1 |
|---|---|
| Molecular Formula | C12H11BO2 |
| Molecular Weight (g/mol) | 198.03 |
| MDL Number | MFCD00093311 |
| InChI Key | XPEIJWZLPWNNOK-UHFFFAOYSA-N |
| PubChem CID | 151253 |
| IUPAC Name | {[1,1'-biphenyl]-4-yl}boronic acid |
| SMILES | OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 5122-94-1 | |
| 198.03 | |
| XPEIJWZLPWNNOK-UHFFFAOYSA-N | |
| {[1,1'-biphenyl]-4-yl}boronic acid |
| C12H11BO2 | |
| MFCD00093311 | |
| 151253 | |
| OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Specifications
| 5122-94-1 | |
| White-Yellow | |
| MFCD00093311 | |
| 4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid | |
| OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
| 198.03 | |
| 198.03 | |
| 4-Biphenylboronic Acid (contains varying amounts of Anhydride) |
| 266°C | |
| C12H11BO2 | |
| 5 g | |
| XPEIJWZLPWNNOK-UHFFFAOYSA-N | |
| {[1,1'-biphenyl]-4-yl}boronic acid | |
| 151253 | |
| Crystalline Powder |
Safety and Handling
TSCA : No