missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 394420050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC394420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5122-94-1
198.03
XPEIJWZLPWNNOK-UHFFFAOYSA-N
151253
B(C1=CC=C(C=C1)C2=CC=CC=C2)(O)O
C12H11BO2
MFCD00093311
4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid
(4-phenylphenyl)boronic acid

Specifications

4-Biphenylboronic acid
96.0
Authentic
Glass bottle
C6H5C6H4B(OH)2
MFCD00093311
XPEIJWZLPWNNOK-UHFFFAOYSA-N
(4-phenylphenyl)boronic acid
151253
97%
5122-94-1
100.0
96% min. (HPLC)
C12H11BO2
5g
4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid
B(C1=CC=C(C=C1)C2=CC=CC=C2)(O)O
198.03
198.03
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

SDS
Documents

Documents

Promotions

Promotions