missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine 98.0+%, TCI America™

Catalog No. B043725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B043725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B043725ML Supplier TCI America Supplier No. B043725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

Melting Point 11°C
Color Yellow
Boiling Point 287°C
Quantity 25 mL
Formula Weight 169.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Benzylpyridine
EINECSNumber (9)-1309
RTECSNumber US2625000
TSCA Yes