missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine, 97%, Thermo Scientific™

Catalog No. AC106160050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106160050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106160050 Supplier Thermo Scientific Chemicals Supplier No. 106160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 9.0°C to 11.0°C
Color Yellow
Density 1.0600g/mL
Boiling Point 287.0°C
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5810 to 1.5830
Linear Formula C6H5CH2(C5H4N)
Quantity 5 mL
Beilstein 20, 426
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents,soluble in ethanol and ether
Formula Weight 169.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Benzylpyridine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-319-0
RTECSNumber US2625000
TSCA TSCA

RUO – Research Use Only